Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

699292

Sigma-Aldrich

trans-2-Phenylvinylboronic acid MIDA ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14BNO4
CAS Number:
Molecular Weight:
259.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

185-195 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)\C=C\c2ccccc2

InChI

1S/C13H14BNO4/c1-15-9-12(16)18-14(19-13(17)10-15)8-7-11-5-3-2-4-6-11/h2-8H,9-10H2,1H3/b8-7+

InChI key

OFLDLTAQNBBGGU-BQYQJAHWSA-N

Application

trans-2-Phenylvinylboronic acid MIDA ester is a stable boronic acid surrogate, which can be used as a reactant in Suzuki-Miyaura cross-coupling reactions . It is also used as a precursor to prepare α-borylated phenyl ketone via the Wacker-type oxidation method.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of ?-borylated ketones by regioselective wacker oxidation of alkenylboronates
Corless VB, et al.
Organic Letters, 20(17), 5300-5303 (2018)
Nicholas A Isley et al.
Journal of the American Chemical Society, 135(47), 17707-17710 (2013-11-15)
New technology has been developed that enables Suzuki-Miyaura couplings involving widely utilized MIDA boronates to be run in water as the only medium, mainly at room temperature. The protocol is such that no organic solvent is involved at any stage;
Panchanan Maiti et al.
International journal of molecular sciences, 21(18) (2020-09-17)
Alzheimer's disease (AD) is characterized by amyloid (Aβ) aggregation, hyperphosphorylated tau, neuroinflammation, and severe memory deficits. Reports that certain boronic compounds can reduce amyloid accumulation and neuroinflammation prompted us to compare trans-2-phenyl-vinyl-boronic-acid-MIDA-ester (TPVA) and trans-beta-styryl-boronic-acid (TBSA) as treatments of deficits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service