Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

699187

Sigma-Aldrich

5-Hexylthiophene-2-carboxaldehyde

97%

Synonym(s):

5-Hexylthiophene-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16OS
CAS Number:
Molecular Weight:
196.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.535

density

1.017 g/mL at 25 °C

SMILES string

CCCCCCc1ccc(C=O)s1

InChI

1S/C11H16OS/c1-2-3-4-5-6-10-7-8-11(9-12)13-10/h7-9H,2-6H2,1H3

InChI key

VWCAHBARARIQLV-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service