Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698997

Sigma-Aldrich

Poly(3-hexylthiophene-2,5-diyl)

regioregular, average Mn 54,000-75,000, electronic grade, 99.995% trace metals basis

Synonym(s):

Plexcore® OS 2100, P3HT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H14S)n
CAS Number:
MDL number:
UNSPSC Code:
12352103

grade

electronic grade

Assay

99.995% trace metals basis

form

solid

mol wt

average Mn 54,000-75,000

description

(>98% head-to-tail regioregular (HNMR))

mp

238 °C

Orbital energy

HOMO 5 eV 
LUMO 3 eV 

OPV Device Performance

ITO/NiO/P3HT/PC61BM/LiF/Al

  • Short-circuit current density (Jsc): 11.3 mA/cm2
  • Open-circuit voltage (Voc): 0.64 V
  • Fill Factor (FF): 0.69
  • Power Conversion Efficiency (PCE): 5.16 %

ITO/PEDOT:PSS/P3HT:PC61BM (1:08)/Al
  • Short-circuit current density (Jsc): 9.5 mA/cm2
  • Open-circuit voltage (Voc): 0.63 V
  • Fill Factor (FF): 0.68
  • Power Conversion Efficiency (PCE): 5 %

Mw/Mn

≤2.5

semiconductor properties

P-type (mobility=10−4 - 10−1 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

General description

LUMO level 3.1 eV
Poly(3-hexylthiophene) (P3HT) is a regioregular semiconducting polymer. It is used in organic electronics primarily because of its regular end-to-end arrangement of side chain, which allows efficient Π-Π stacking of the conjugated backbones. On account of the alkyl side group, P3HT is rendered hydrophobic in neutral state.[1]

Application

High molecular weight; ultra-high purity P3HT optimized for use in organic photovoltaics (OPV) research and devices. Poly(3-hexylthiophene-2,5-diyl) may be used extensively as a semiconducting layer in organic thin film field effect transistor (FETs) and solar cells.[2],2

Legal Information

Product of Plextronics, Inc. U.S. Patent 6,166,172.
Plexcore is a registered trademark of Plextronics, Inc.

comparable product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Control of active semiconducting layer packing in organic thin film transistors through synthetic tailoring of dielectric materials
Singh R, et al.
Royal Society of Chemistry Advances, 4(56), 29383-29392 (2014)
High sensitivity organic photodiodes with low dark currents and increased lifetimes
Ramuz, M.; Burgi, L.; Winnewisser, C.; et al.
Organic Electronics, 9, 369-376 (2008)
Lei Yang et al.
Physical chemistry chemical physics : PCCP, 14(2), 779-789 (2011-11-26)
Two hole conductor materials, spiro-OMeTAD and P3HT, were compared in solid-state dye-sensitized solar cells. Two organic dyes containing one anchor unit (D35) or two anchor units (M3) were used in the comparison. Absorbed photon to current conversion efficiency close to
Thermally Stable, Efficient Polymer Solar Cells with Nanoscale Control of the Interpenetrating Network Morphology
Ma, W.; et al.
Advances in Functional Materials, 15, 1617-1622 (2005)
Flexible Transistors Exploiting P3HT on Paper Substrate and Graphene Oxide Film as Gate Dielectric: Proof of Concept.
Valentini L, et al.
Science of Advanced Materials, 5(5), 530-533 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service