Skip to Content
MilliporeSigma
All Photos(1)

Documents

698709

Sigma-Aldrich

Allylboronic acid MIDA ester

Synonym(s):

2-Allyl-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12BNO4
Molecular Weight:
197.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

CN1CC(=O)OB(CC=C)OC(=O)C1

InChI

1S/C8H12BNO4/c1-3-4-9-13-7(11)5-10(2)6-8(12)14-9/h3H,1,4-6H2,2H3

InChI key

RKMASTMFHKLQNL-UHFFFAOYSA-N

Other Notes

May contain up to 10 wt. % dimethyl sulfoxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabin Llona-Minguez et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(20), 7394-7398 (2015-03-27)
A two-step synthesis of structurally diverse pyrrole-containing bicyclic systems is reported. ortho-Nitro-haloarenes coupled with vinylic N-methyliminodiacetic acid (MIDA) boronates generate ortho-vinyl-nitroarenes, which undergo a "metal-free" nitrene insertion, resulting in a new pyrrole ring. This novel synthetic approach has a wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service