Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69796

Sigma-Aldrich

(+)-Ethyl D-lactate

≥99.0% (sum of enantiomers, GC)

Synonym(s):

(+)-Ethyl (R)-2-hydroxypropionate, (R)-(+)-2-Hydroxypropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1720840
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, GC)

form

liquid

optical purity

enantiomeric ratio: ≥99:1 (GC)

SMILES string

CCOC(=O)[C@@H](C)O

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m1/s1

InChI key

LZCLXQDLBQLTDK-SCSAIBSYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Sun et al.
Journal of environmental quality, 38(4), 1591-1597 (2009-06-25)
A new approach using aqueous ethyl lactate-modified [S,S]-ethylenediaminedisuccinic acid (EDDS) washing solutions was examined in the laboratory for the simultaneous removal of phenanthrene, pyrene, and Cu from contaminated soils. Ethyl lactate demonstrated greater solubilization efficiency for phenanthrene and pyrene than
Chiaki Inaba et al.
Applied microbiology and biotechnology, 83(5), 859-864 (2009-03-17)
The whole-cell biocatalyst displaying Candida antarctica lipase B (CALB) on the yeast cell surface with alpha-agglutinin as the anchor protein was easy to handle and possessed high stability. The lyophilized CALB-displaying yeasts showed their original hydrolytic activity and were applied
Jun Liu et al.
The Journal of organic chemistry, 77(21), 9718-9723 (2012-10-12)
The efficient and concise synthesis of (-)-orthodiffenes A and C has been accomplished for the first time in eight steps from readily available chiral synthons, D-mannose and D-ethyl lactate. Our work confirmed the complete structure of orthodiffenes A and C
Jayashree Jayaraj et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 39(6), 1399-1414 (2004-07-13)
Chloroethenes like tetrachloroethene (PCE) are the most prevalent groundwater contaminants in the USA. Their presence as nonaqueous phase liquids (NAPLs) makes remediation difficult. Among options for NAPL cleanup, co-solvent injection has demonstrated success. However, the process has the potential to
Jiahua Li et al.
Journal of hazardous materials, 181(1-3), 673-678 (2010-06-18)
In order to explore a practical approach to the remediation of the cadmium (Cd)-contaminated soil in the lower reaches of the Yangtze River, we evaluated the effects of a local willow (Salix x aureo-pendula CL 'J1011') of absorbing, accumulating, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service