Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697893

Sigma-Aldrich

6-Bromoquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6BrN
CAS Number:
Molecular Weight:
208.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.663

density

1.538 g/mL at 25 °C

functional group

bromo

SMILES string

Brc1ccc2ncccc2c1

InChI

1S/C9H6BrN/c10-8-3-4-9-7(6-8)2-1-5-11-9/h1-6H

InChI key

IFIHYLCUKYCKRH-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alexander V Shaferov et al.
Sensors (Basel, Switzerland), 20(11) (2020-06-11)
Pd(0)-catalyzed amination was employed for the synthesis of a new family of (S)-1,1'-bianaphthalene-2,2'-diamine derivatives possessing additional chiral and fluorophore substituents. The compounds thus obtained were tested as potential detectors of seven amino alcohols, and some of them were found to
Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service