Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697613

Sigma-Aldrich

Phenylsulfur pentafluoride

Synonym(s):

Pentafluoro(phenyl)sulfur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F5S
CAS Number:
Molecular Weight:
204.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.435

density

1.496 g/mL at 25 °C

SMILES string

FS(F)(F)(F)(F)c1ccccc1

InChI

1S/C6H5F5S/c7-12(8,9,10,11)6-4-2-1-3-5-6/h1-5H

InChI key

DMNYFEWFSFTYEL-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.9 °F

Flash Point(C)

43.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service