Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697605

Sigma-Aldrich

(S,S)-ANDEN-Phenyl Trost Ligand

Synonym(s):

(+)-(11S,12S)Bis[2′-(diphenylphosphino)benzamido]-9,10-dihydro-9,10-ethanoanthracene, (11S,12S)-N,N′-(9,10-Dihydro-9,10-ethanoanthracene-11,12-diyl)bis[2-(diphenylphosphino)]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H42N2O2P2
CAS Number:
Molecular Weight:
812.87
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

optical activity

[α]22/D +111°, c = 1 in chloroform

mp

123-128 °C

SMILES string

O=C(N[C@@H]1[C@@H](NC(=O)c2ccccc2P(c3ccccc3)c4ccccc4)[C@H]5c6ccccc6[C@@H]1c7ccccc57)c8ccccc8P(c9ccccc9)c%10ccccc%10

InChI

1S/C54H42N2O2P2/c57-53(45-33-17-19-35-47(45)59(37-21-5-1-6-22-37)38-23-7-2-8-24-38)55-51-49-41-29-13-15-31-43(41)50(44-32-16-14-30-42(44)49)52(51)56-54(58)46-34-18-20-36-48(46)60(39-25-9-3-10-26-39)40-27-11-4-12-28-40/h1-36,49-52H,(H,55,57)(H,56,58)/t49-,50+,51-,52-/m0/s1

InChI key

BEKZNCOFLFMFHP-UYATXGGZSA-N

Application

Asymmetric allylic alkylation ligand.

Legal Information

Sold in collaboration with Dr. Reddy′s Laboratories for research purposes only.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trost, B.M. et al.
Aldrichimica Acta, 40, 59-59 (2007)
Trost, B.M. et al.
Journal of the American Chemical Society, 124, 11616-11616 (2004)
Asymmetric Transition Metal-Catalyzed Allylic Alkylations.
Barry M. Trost et al.
Chemical reviews, 96(1), 395-422 (1996-02-01)
Asymmetric transition-metal-catalyzed allylic alkylations: applications in total synthesis.
Barry M Trost et al.
Chemical reviews, 103(8), 2921-2944 (2003-08-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service