Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697532

Sigma-Aldrich

2-(Dicyclohexylphosphino)benzophenone

95%

Synonym(s):

[2-(Dicyclohexylphosphino)phenyl]phenylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31OP
CAS Number:
Molecular Weight:
378.49
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

139-146 °C

functional group

phosphine

SMILES string

O=C(c1ccccc1)c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C25H31OP/c26-25(20-12-4-1-5-13-20)23-18-10-11-19-24(23)27(21-14-6-2-7-15-21)22-16-8-3-9-17-22/h1,4-5,10-13,18-19,21-22H,2-3,6-9,14-17H2

InChI key

NFAAYQZZHNIMDJ-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service