Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697370

Sigma-Aldrich

2-[(11bS)-3H-Binaphtho[2,1-c:1′,2′-e]phosphepin-4(5H)-yl]ethanamine

kanata purity

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22NP
Molecular Weight:
355.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

powder

mp

64-70 °C

SMILES string

NCCP1Cc2ccc3ccccc3c2-c4c(C1)ccc5ccccc45

InChI

1S/C24H22NP/c25-13-14-26-15-19-11-9-17-5-1-3-7-21(17)23(19)24-20(16-26)12-10-18-6-2-4-8-22(18)24/h1-12H,13-16,25H2

InChI key

PXDLCEFIOZLJLW-UHFFFAOYSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service