Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697354

Sigma-Aldrich

(Rp)-1-[(1S)-(1-Aminoethyl)]-2-(diphenylphosphino)ferrocene

97%

Synonym(s):

(Rp)-1-(1S)-[(2-Diphenylphosphino)ferrocenyl]ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24FeNP
CAS Number:
Molecular Weight:
413.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

131-135 °C

functional group

amine
phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H](N)[C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4

InChI

1S/C19H19NP.C5H5.Fe/c1-15(20)18-13-8-14-19(18)21(16-9-4-2-5-10-16)17-11-6-3-7-12-17;1-2-4-5-3-1;/h2-15H,20H2,1H3;1-5H;/t15-;;/m0../s1

InChI key

DRBVPPSGNFMTKZ-CKUXDGONSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Aminophosphine Ligands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service