Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696323

Sigma-Aldrich

Sodium DL-3-hydroxybutyrate-1-13C

98 atom % 13C, ≥99% (CP)

Synonym(s):

DL-3-Hydroxybutyric acid-1-13C sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH213CO2Na
CAS Number:
Molecular Weight:
127.08
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % 13C

Assay

≥99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

161-167 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

[Na+].CC(O)C[13C]([O-])=O

InChI

1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/i4+1;

InChI key

NBPUSGBJDWCHKC-VZHAHHFWSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service