Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696242

Sigma-Aldrich

Bis[1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]copper(I) tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H48BCuF4N4
Molecular Weight:
759.21
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

reagent type: catalyst
core: copper

SMILES string

F[B-](F)(F)F.Cc1cc(C)c(N2C=CN(C2[Cu+]C3N(C=CN3c4c(C)cc(C)cc4C)c5c(C)cc(C)cc5C)c6c(C)cc(C)cc6C)c(C)c1

InChI

1S/2C21H25N2.BF4.Cu/c2*1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;2-1(3,4)5;/h2*7-13H,1-6H3;;/q;;-1;+1

InChI key

KYKUFSRBHPLMIP-UHFFFAOYSA-N

Application

Efficient precatalyst for hydrosililation of carbonyl compounds.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service