Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696099

Sigma-Aldrich

2,3-Diaminobenzoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

95%

mp

198-204 °C

application(s)

peptide synthesis

SMILES string

Nc1cccc(C(O)=O)c1N

InChI

1S/C7H8N2O2/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3H,8-9H2,(H,10,11)

InChI key

KKTUQAYCCLMNOA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geun Woo Dong et al.
Clinical and experimental otorhinolaryngology, 2(1), 13-19 (2009-05-13)
Despite the ongoing development of treatment protocols for laryngeal squamous cell carcinoma (LSCC), the patients suffering with this malady have shown only a modestly improved outcome. This poor outcome has been attributed to the lack of therapy that's individualized to
Takeshi Matsumoto et al.
Scientific reports, 8(1), 14623-14623 (2018-10-04)
We report the photochemical carboxylation of o-phenylenedimamine in the absence of a base and an electron donor under an atmosphere of CO2, which afforded 2,3-diaminobenzoic acid (DBA) in 28% synthetic yield and 0.22% quantum yield (Φ(%)). The synthetic yield of
Anal Jana et al.
Journal of photochemistry and photobiology. B, Biology, 214, 112091-112091 (2020-12-08)
Formaldehyde (FA), a simple reactive carbonyl molecule, is endogenously produced in the cell at various physiological condition. At elevated level, FA causes severe cell toxicity as well as damage in macromolecules such proteins and DNA. For detecting FA in living
Bettina S Buchmaier et al.
PloS one, 8(7), e68301-e68301 (2013-07-23)
Osmotic stress has been shown to regulate cytoskeletal protein expression. It is generally known that vimentin is rapidly degraded during apoptosis by multiple caspases, resulting in diverse vimentin fragments. Despite the existence of the known apoptotic vimentin fragments, we demonstrated
Alexander von Tesmar et al.
Cell chemical biology, 24(10), 1216-1227 (2017-09-12)
In vitro reconstitution and biochemical analysis of natural product biosynthetic pathways remains a challenging endeavor, especially if megaenzymes of the nonribosomal peptide synthetase (NRPS) type are involved. In theory, all biosynthetic steps may be deciphered using mass spectrometry (MS)-based analyses of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service