Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695998

Sigma-Aldrich

Hexamethylenetetramine-13C6, 15N4

99 atom % 13C, 98 atom % 15N, 97% (CP)

Synonym(s):

Hexamine-13C6,15N4, Methenamine-13C6,15N4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H1215N4
CAS Number:
Molecular Weight:
150.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

97% (CP)

form

solid

mp

280 °C (subl) (lit.)

mass shift

M+10

SMILES string

[13CH2]1[15N]2[13CH2][15N]3[13CH2][15N]1[13CH2][15N]([13CH2]2)[13CH2]3

InChI

1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1

InChI key

VKYKSIONXSXAKP-IIYFYTTLSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2 - Resp. Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service