Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695750

Sigma-Aldrich

Ethyl (5-bromo-2-fluorobenzoyl)acetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BrFO3
CAS Number:
Molecular Weight:
289.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

powder

mp

34-39 °C

functional group

bromo
ester
fluoro
ketone

SMILES string

CCOC(=O)CC(=O)c1cc(Br)ccc1F

InChI

1S/C11H10BrFO3/c1-2-16-11(15)6-10(14)8-5-7(12)3-4-9(8)13/h3-5H,2,6H2,1H3

InChI key

XODIHCQWLRSGQI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service