Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695181

Sigma-Aldrich

Bis(2,2-dimethylpropanoato)(4-methylphenyl)bis[tris[4-(trifluoromethyl)phenyl]phosphine]rhodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H49F18O4P2Rh
CAS Number:
Molecular Weight:
1328.84
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

mp

204-206 °C

SMILES string

Cc1ccc(cc1)[Rh](OC(=O)C(C)(C)C)OC(=O)C(C)(C)C.FC(F)(F)c2ccc(cc2)P(c3ccc(cc3)C(F)(F)F)c4ccc(cc4)C(F)(F)F.FC(F)(F)c5ccc(cc5)P(c6ccc(cc6)C(F)(F)F)c7ccc(cc7)C(F)(F)F

InChI

1S/2C21H12F9P.C7H7.2C5H10O2.Rh/c2*22-19(23,24)13-1-7-16(8-2-13)31(17-9-3-14(4-10-17)20(25,26)27)18-11-5-15(6-12-18)21(28,29)30;1-7-5-3-2-4-6-7;2*1-5(2,3)4(6)7;/h2*1-12H;3-6H,1H3;2*1-3H3,(H,6,7);/q;;;;;+2/p-2

InChI key

XAQKQHIYPBKBNW-UHFFFAOYSA-L

General description

Extremely selective C-arylation catalyst for unprotected indoles and pyrrolidines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service