Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695165

Sigma-Aldrich

Bis(3,5-dimethylphenyl)chlorophosphine

Synonym(s):

Bis(3,5-dimethylphenyl)phosphinous chloride, Chlorobis(3,5-dimethylphenyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClP
CAS Number:
Molecular Weight:
276.74
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.606

density

1.102 g/mL at 25 °C

functional group

phosphine

SMILES string

Cc1cc(C)cc(c1)P(Cl)c2cc(C)cc(C)c2

InChI

1S/C16H18ClP/c1-11-5-12(2)8-15(7-11)18(17)16-9-13(3)6-14(4)10-16/h5-10H,1-4H3

InChI key

FCEBDAANWYNQMO-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Ocansey et al.
Frontiers in chemistry, 8, 591353-591353 (2020-12-12)
A rise in atmospheric CO2 levels, following years of burning fossil fuels, has brought about increase in global temperatures and climate change due to the greenhouse effect. As such, recent efforts in addressing this problem have been directed to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service