Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694738

Sigma-Aldrich

2-Amino-3-iodo-5-nitropyridine

97%

Synonym(s):

3-Iodo-5-nitro-2-pyridinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4IN3O2
CAS Number:
Molecular Weight:
265.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

231-236 °C

functional group

iodo
nitro

SMILES string

Nc1ncc(cc1I)[N+]([O-])=O

InChI

1S/C5H4IN3O2/c6-4-1-3(9(10)11)2-8-5(4)7/h1-2H,(H2,7,8)

InChI key

FEADVZZIWYNCBF-UHFFFAOYSA-N

Application

Useful starter in azaindole chemistry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isao Takemura et al.
Organic letters, 6(15), 2503-2505 (2004-07-17)
[reaction: see text] Concise synthesis of defucogilvocarcin M was achieved via the [2 + 2 + 2] approach to beta-phenylnaphthalene structure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service