Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694363

Sigma-Aldrich

Methyl 1-(trifluoromethyl)cyclopropane-1-carboxylate

95%

Synonym(s):

1-(Trifluoromethyl)cyclopropane-1-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7F3O2
CAS Number:
Molecular Weight:
168.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.373

density

1.241 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COC(=O)C1(CC1)C(F)(F)F

InChI

1S/C6H7F3O2/c1-11-4(10)5(2-3-5)6(7,8)9/h2-3H2,1H3

InChI key

PAWUSHZXEZPDBE-UHFFFAOYSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

90.0 °F

Flash Point(C)

32.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service