Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694096

Sigma-Aldrich

2-Butynyl p-toluenesulfonate

96%

Synonym(s):

2-Butynyl 4-methylbenzenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3S
CAS Number:
Molecular Weight:
224.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

47-51 °C

SMILES string

CC#CCOS(=O)(=O)c1ccc(C)cc1

InChI

1S/C11H12O3S/c1-3-4-9-14-15(12,13)11-7-5-10(2)6-8-11/h5-8H,9H2,1-2H3

InChI key

HGZKJNHJABSJTC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad S Rahnama'i et al.
BMC urology, 13, 8-8 (2013-02-08)
To investigate the effect of prostaglandin depletion by means of COX-inhibition on cholinergic enhanced spontaneous contractions. The urethra and bladder of 9 male guinea pigs (weight 270-300 g) were removed and placed in an organ bath with Krebs' solution. A
Yuji Odagaki et al.
Journal of pharmacological sciences, 125(2), 157-168 (2014-05-23)
The functional activation of Gi/o proteins coupled to muscarinic acetylcholine receptors (mAChRs) was investigated with the conventional guanosine-5'-O-(3-[(35)S]thio) triphosphate ([(35)S]GTPγS) binding assay in rat brain membranes. The most efficacious stimulation elicited by acetylcholine or carbachol (CCh) was obtained in striatal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service