Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693979

Sigma-Aldrich

(1E,2E)-1,2-Bis(2,6-Diisopropylphenylimino)ethane

90%

Synonym(s):

N,N′-Bis(2,6-diisopropylphenyl)-1,4-diazabutadiene, N,N′-Bis(2,6-diisopropylphenyl)ethanediimine, Glyoxal bis(2,6-diisopropylanil)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H7)2C6H3NCHCHNC6H3(C3H7)2
CAS Number:
Molecular Weight:
376.58
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

reaction suitability

reagent type: ligand

mp

105-109 °C

SMILES string

CC(C)c1cccc(C(C)C)c1\N=C\C=N\c2c(cccc2C(C)C)C(C)C

InChI

1S/C26H36N2/c1-17(2)21-11-9-12-22(18(3)4)25(21)27-15-16-28-26-23(19(5)6)13-10-14-24(26)20(7)8/h9-20H,1-8H3/b27-15+,28-16+

InChI key

JWVIIGXMTONOFR-DPCVLPDWSA-N

Application

  • Reactant in preparation of derived ruthenium olefin metathesis catalysts
  • N-cyclic carbene ligand

Catalyst in:
  • Palladium-catalyzed aerobic alc. oxidn. supported by a-diimine ligands
  • Regioselective alkylation in presence of ruthenium-bisimine catalytic precursors
  • N-arylation of aromatic amines
  • Preparation of ruthenium nitrosyl alpha-diimine and iminoketone complexes as catalysts for transfer hydrogenation of ketones and atom transfer radical polymerization reactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service