Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

693928

Sigma-Aldrich

Yttrium-Nickel alloy, YNi5

greener alternative

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YNi5
CAS Number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Ni].[Ni].[Ni].[Ni].[Ni].[Y]

InChI

1S/5Ni.Y

InChI key

PKGOVESIHDGGOX-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service