Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693820

Sigma-Aldrich

Neodymium-iron-boron, alloy 30/200

disc, size 13 mm × 6 mm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

disc

composition

Aluminum, 0.8 wt. %
Boron, 1.1 wt. %
Dysprosium, 3.6 wt. %
Iron, 65 wt. %
Neodymium, 29 wt. %
Niobium, 0.5 wt. %

size

13 mm × 6 mm

SMILES string

[B].[Nd].[Fe]

InChI

1S/B.Fe.Nd

InChI key

QJVKUMXDEUEQLH-UHFFFAOYSA-N

Other Notes

Magnetic parameters/properties:
Maximum Energy Product (BH): 248 kJ/m3 (31 MGsOe)
Residual Induction (Br): 1.14 T (11.4 kGs)
Coercive Force (Hc): 835 kA/m (10.5 kOe)
Intrinsic Coercive Force (Hci): 2400 kA/m (30.0 kOe)
Maximum Temperature rate: 200°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service