Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

693618

Sigma-Aldrich

(2S,4S)-2,4-Bis(diphenylphosphino)pentane

97%

Synonym(s):

(S,S)-BDPP, [(1S,3S)-1,3-Dimethyl-1,3-propanediyl]bis[diphenylphosphine]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H30P2
CAS Number:
Molecular Weight:
440.50
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D −125.0°, c = 1 in chloroform

mp

80-84 °C

functional group

phosphine

SMILES string

C[C@@H](C[C@H](C)P(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C29H30P2/c1-24(30(26-15-7-3-8-16-26)27-17-9-4-10-18-27)23-25(2)31(28-19-11-5-12-20-28)29-21-13-6-14-22-29/h3-22,24-25H,23H2,1-2H3/t24-,25-/m0/s1

InChI key

CTYPJIUQROQJBG-DQEYMECFSA-N

Application

Ligand used for asymmetric hydrogenation of ketones[1] and alkenes[2]

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideo Shimizu et al.
Organic letters, 9(9), 1655-1657 (2007-03-29)
[reaction: see text] A novel asymmetric hydrogenation protocol using a copper catalyst is reported. A Cu(I) complex in the presence of nonracemic BDPP hydrogenates aryl ketones with moderate to high enantioselectivity.
Shui-Ming Lu et al.
Journal of the American Chemical Society, 130(20), 6451-6455 (2008-05-01)
The intramolecular cyclocarbonylation reaction with palladium-complexed dendrimers on silica is a very effective method for the regioselective synthesis of methylene 8-, 9-, and 10-membered rings. The heterogeneous dendritic catalysts are easily recovered by simple filtration and reused for up to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service