Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693421

Sigma-Aldrich

3-[(E)-[[(1R,2R)-2-[[(1S)-1-[(Dimethylamino)carbonyl]-2,2-dimethylpropyl]thioureido]cyclohexyl]imino]methyl]-5-(tert-butyl)-4-hydroxyphenyl pivalate

95%

Synonym(s):

3-[[[(1R,2R)-2-[[[[(1S)-1-[(Dimethylamino)carbonyl]-2,2-dimethylpropyl]amino]thioxomethyl]amino]cyclohexyl]imino]methyl]-5-(1,1-dimethylethyl)-4-hydroxyphenyl 2,2-dimethylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H50N4O4S
CAS Number:
Molecular Weight:
574.82
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

95%

optical activity

[α]20/D -88°, c = 1 in chloroform

mp

135-138 °C (D)

Mw/Mn

<1.2

SMILES string

CN(C)C(=O)[C@@H](NC(=S)N[C@@H]1CCCC[C@H]1\N=C\c2cc(OC(=O)C(C)(C)C)cc(c2O)C(C)(C)C)C(C)(C)C

InChI

1S/C31H50N4O4S/c1-29(2,3)21-17-20(39-27(38)31(7,8)9)16-19(24(21)36)18-32-22-14-12-13-15-23(22)33-28(40)34-25(30(4,5)6)26(37)35(10)11/h16-18,22-23,25,36H,12-15H2,1-11H3,(H2,33,34,40)/b32-18+/t22-,23-,25-/m1/s1

InChI key

BOFCLIFENISFHX-ZIKUWVHDSA-N

Application

Catalyst for the Strecker reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petr Vachal et al.
Journal of the American Chemical Society, 124(34), 10012-10014 (2002-08-22)
A mechanistic investigation of the asymmetric Strecker reaction catalyzed by a metal-free Schiff base catalyst was conducted. The active site of the catalyst, the relevant stereoisomer of the imine substrate, and the solution structure of the imine-catalyst complex were elucidated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service