Skip to Content
MilliporeSigma
All Photos(1)

Documents

693413

Sigma-Aldrich

(S)-2-[[(1R,2R)-2-Aminocyclohexyl]thioureido]-N-benzyl-N,3,3-trimethylbutanamide

95%

Synonym(s):

(2S)-2-[[[[(1R,2R)-2-Aminocyclohexyl]amino]thioxomethyl]amino]-N-3,3-trimethyl-N-(phenylmethyl)butanamide, (S)-2-[[[[(1R,2R)-2-Aminocyclohexyl]amino]thioxomethyl]amino]-N-benzyl-N-3,3-trimethylbutanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34N4OS
CAS Number:
Molecular Weight:
390.59
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

95%

optical activity

[α]20/D +19.0°, c = 1 in chloroform

mp

71-85 °C

SMILES string

CN(Cc1ccccc1)C(=O)[C@@H](NC(=S)N[C@@H]2CCCC[C@H]2N)C(C)(C)C

InChI

1S/C21H34N4OS/c1-21(2,3)18(19(26)25(4)14-15-10-6-5-7-11-15)24-20(27)23-17-13-9-8-12-16(17)22/h5-7,10-11,16-18H,8-9,12-14,22H2,1-4H3,(H2,23,24,27)/t16-,17-,18-/m1/s1

InChI key

CNEVMNSDRMDKFO-KZNAEPCWSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service