Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693391

Sigma-Aldrich

Poly(3-decyloxythiophene-2,5-diyl)

0.5 % (w/v) in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C14H22OS)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid

mol wt

Mn <10,000

concentration

0.5 % (w/v) in THF

density

0.877 g/mL at 25 °C

fluorescence

λex 350 nm; λem 427 nm in THF

General description

Regioregular.[1]

Application

P3DOT may be used in solar cells.1,[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.9 °F

Flash Point(C)

-23.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenjun Shi et al.
Journal of the American Chemical Society, 128(27), 8980-8986 (2006-07-06)
Low band gap conjugated polymers with proper energy levels for charge transfer are required to achieve high-efficiency polymer solar cells. We report the synthesis and characterization of two new regioregular copolymers that are based on 3-alkoxythiophene monomers: poly(3-octylthiophene-2,5-diyl-co-3-decyloxythiophene-2,5-diyl) (POT-co-DOT) and

Articles

Materials Design Concepts for Efficient Blue OLEDs: A Joint Theoretical and Experimental Study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service