Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692859

Sigma-Aldrich

Samarium-cobalt alloy 18

disc, size 10 mm × 6 mm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SmCo5
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

disc

composition

Cobalt, 62.75 wt. %
Samarium, 37.25 wt. %

size

10 mm × 6 mm

InChI

1S/Co.Sm

InChI key

KPLQYGBQNPPQGA-UHFFFAOYSA-N

Linkage

For additional technical details, view the Materials Science Technology Spotlight on magnetic alloys at www.sigma-aldrich.com/magnetic

Other Notes

Magnetic parameters/properties:
Maximum Energy Product (BH): 140 kJ/m3 (18 MGsOe)
Residual Induction (Br): 0.87 T (8.7 kGs)
Coercive Force (Hc): 680 kA/m (8.5 kOe)
Intrinsic Coercive Force (Hci): 2390 kA/m (30.0 kOe)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Solid state and materials chemistry have made a tremendous impact and have experienced growth in recent years, particularly for rare earthcontaining materials.

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service