Skip to Content
MilliporeSigma
All Photos(1)

Documents

692840

Sigma-Aldrich

Samarium-cobalt alloy 24

disc, size 10 mm × 6 mm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm2Co17
MDL number:
UNSPSC Code:
11101711
PubChem Substance ID:
NACRES:
NA.23

form

disc

composition

Cobalt, 49.05 wt. %
copper, 5.95 wt. %
Iron, 17.25 wt. %
Samarium, 25 wt. %
Zirconium, 2.75 wt. %

size

10 mm × 6 mm

InChI

1S/Co.Sm

InChI key

KPLQYGBQNPPQGA-UHFFFAOYSA-N

Other Notes

Magnetic parameters/properties:
Maximum Energy Product (BH): 190 kJ/m3 (24 MGsOe)
Residual Induction (Br): 1.00 T (10.0 kGs)
Coercive Force (Hc): 740 kA/m (9.3 kOe)
Intrinsic Coercive Force (Hci): 2070 kA/m (26.0 kOe)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Solid state and materials chemistry have made a tremendous impact and have experienced growth in recent years, particularly for rare earthcontaining materials.

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service