Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692468

Sigma-Aldrich

(R)-[(RuCl(DM-BINAP))2(μ-Cl)3][NH2Me2]

Synonym(s):

Dimethylammonium dichlorotri(μ-chloro)bis[(R)-(+)-2,2′-bis[di(3,5-xylyl)phosphino]-1,1′-binaphthyl]diruthenate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH2+[C104H96Cl5P4Ru2]-
CAS Number:
Molecular Weight:
1895.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

storage temp.

2-8°C

SMILES string

Cl.[Cl-].CNC.[Cl+]1[Ru-]2[Cl+][Ru-]1[Cl+]2.Cc3cc(C)cc(c3)P(c4cc(C)cc(C)c4)c5ccc6ccccc6c5-c7c(ccc8ccccc78)P(c9cc(C)cc(C)c9)c%10cc(C)cc(C)c%10.Cc%11cc(C)cc(c%11)P(c%12cc(C)cc(C)c%12)c%13ccc%14ccccc%14c%13-c%15c(ccc%16ccccc%15%16)P(c%17cc(C)cc(C)c%17)c%18cc(C)cc(C)c%18

InChI

1S/2C52H48P2.C2H7N.2ClH.3Cl.2Ru/c2*1-33-21-34(2)26-43(25-33)53(44-27-35(3)22-36(4)28-44)49-19-17-41-13-9-11-15-47(41)51(49)52-48-16-12-10-14-42(48)18-20-50(52)54(45-29-37(5)23-38(6)30-45)46-31-39(7)24-40(8)32-46;1-3-2;;;;;;;/h2*9-32H,1-8H3;3H,1-2H3;2*1H;;;;;/q;;;;;3*+1;2*-1/p-1

InChI key

LGSCCUXQQTYWAH-UHFFFAOYSA-M

Legal Information

Sold in collaboration with Takasago for reseach purposes only. JP Registration No. 3445074

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service