Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692344

Sigma-Aldrich

DPBP-bidentate phosphine

95%

Synonym(s):

2,2′ Bis(diphenylphosphino)benzophenone, Bis[2-(diphenylphosphino)phenyl]methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H28OP2
CAS Number:
Molecular Weight:
550.57
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

152-156 °C

functional group

phosphine

SMILES string

O=C(c1ccccc1P(c2ccccc2)c3ccccc3)c4ccccc4P(c5ccccc5)c6ccccc6

InChI

1S/C37H28OP2/c38-37(33-25-13-15-27-35(33)39(29-17-5-1-6-18-29)30-19-7-2-8-20-30)34-26-14-16-28-36(34)40(31-21-9-3-10-22-31)32-23-11-4-12-24-32/h1-28H

InChI key

RKNMFQXZZRUBQH-UHFFFAOYSA-N

Application

Reactant for benzophenone-derived catalysts

Legal Information

Sold in collaboration with Takasago for research purposes only. US7473793

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service