Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691968

Sigma-Aldrich

Tris(2-chloroethyl) phosphite

85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CH2O)3P
CAS Number:
Molecular Weight:
269.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

85%

refractive index

n20/D 1.487 (lit.)

bp

112-115 °C/2 mmHg (lit.)

density

1.328 g/mL at 25 °C (lit.)

SMILES string

ClCCOP(OCCCl)OCCCl

InChI

1S/C6H12Cl3O3P/c7-1-4-10-13(11-5-2-8)12-6-3-9/h1-6H2

InChI key

LUVCTYHBTXSAMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

374.9 °F

Flash Point(C)

190.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service