Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691763

Sigma-Aldrich

Azelaic acid-13C9

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOO13C(13CH2)713COOH
Molecular Weight:
197.15
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

mp

98-103 °C (lit.)

mass shift

M+9

SMILES string

O[13C](=O)[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1

InChI key

BDJRBEYXGGNYIS-RIQGLZQJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact

Other Notes

Bulk stock may be available for this product. Please contact the Stable Isotope Customer Service Group at 937-859-1808 (1-800-448-9760 for US & Canada) or email us at [email protected]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service