Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

691704

Sigma-Aldrich

Hexadecahydropyrene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26
CAS Number:
Molecular Weight:
218.38
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.521

density

0.984 g/mL at 25 °C

SMILES string

C1CC2CCC3CCCC4CCC(C1)C2C34

InChI

1S/C16H26/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h11-16H,1-10H2

InChI key

BYBPEZLZCGOWIS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Letinski et al.
Chemosphere, 150, 416-423 (2016-03-01)
Aqueous solubility is a fundamental physical-chemical substance property that strongly influences the distribution, fate and effects of chemicals upon release into the environment. Experimental water solubility was determined for 18 selected C9-C18 normal, branched and cyclic alkanes. A slow-stir technique

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service