Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691631

Sigma-Aldrich

3,3′′′-Didodecyl-2,2′:5′,2′′:5′′,2′′′−quaterthiophene

97%

Synonym(s):

5,5′-Bis(3-dodecyl-2-thienyl)-2,2′-bithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H58S4
CAS Number:
Molecular Weight:
667.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

55-59 °C

SMILES string

CCCCCCCCCCCCc1ccsc1-c2ccc(s2)-c3ccc(s3)-c4sccc4CCCCCCCCCCCC

InChI

1S/C40H58S4/c1-3-5-7-9-11-13-15-17-19-21-23-33-29-31-41-39(33)37-27-25-35(43-37)36-26-28-38(44-36)40-34(30-32-42-40)24-22-20-18-16-14-12-10-8-6-4-2/h25-32H,3-24H2,1-2H3

InChI key

DZUNDTRLGXGTGU-UHFFFAOYSA-N

Application

Semiconducting oligomer (p-type), also used as monomer to prepare poly(3,3′′′-dialkyl-quaterthiophene) polymers (PQTs).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beng S Ong et al.
Journal of the American Chemical Society, 126(11), 3378-3379 (2004-03-18)
Conjugated polymers have been widely studied as potential semiconductor materials for organic thin-film transistors (TFTs). However, they have provided functionally poor transistor properties when the TFTs are fabricated in air. We have developed a class of liquid crystalline regioregular polythiophenes

Articles

Review the potential of self-assembled multilayer gate dielectric films fabricated from silane precursors for organic, inorganic, and transparent TFT and for TFT circuitry and OLED displays.

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service