Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691267

Sigma-Aldrich

1,3-Dimethoxy-2-methylimidazolium bis(trifluoromethylsulfonyl)imide

98%, complies for IR spectroscopy

Synonym(s):

(OMe)2MeIm-NTf2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11F6N3O6S2
CAS Number:
Molecular Weight:
423.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (13C-NMR)
≥98% (19F-NMR)
≥98% (31P-NMR)
98%

form

liquid

impurities

1.1 % (w/w) PF6 (19F-NMR)

mp

58 °C

suitability

complies for IR spectroscopy

functional group

fluoro

SMILES string

COn1cc[n+](OC)c1C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H11N2O2.C2F6NO4S2/c1-6-7(9-2)4-5-8(6)10-3;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h4-5H,1-3H3;/q+1;-1

InChI key

YBRWKTHXZKFEDT-UHFFFAOYSA-N

Application

1,3-Dimethoxy-2-methylimidazolium bis(trifluoromethylsulfonyl)imide can be used as:
  • An ionic liquid and NHC precatalyst for cross-coupling reactions.
  • A solvent in the study of solid-liquid equilibria (SLE) of brown coal.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-liquid equilibria predictions for the dissolution of brown coal in ionic liquids using a continuum solvation model
Bhoi S, et al.
Fuel Processing Technology, 126, 112-121 (2014)
N,N-Di(alkoxy)imidazolium salts: new patent-free ionic liquids and NHC-precatalysts
Laus, G., et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 62, 295-308 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service