Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690619

Sigma-Aldrich

2,6-Bis[1-(2-methylphenylimino)ethyl]pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23N3
CAS Number:
Molecular Weight:
341.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96.5-103.5% (titration)
97%

form

crystals

reaction suitability

reagent type: ligand

SMILES string

Cc1ccccc1\N=C(/C)c2cccc(n2)\C(C)=N\c3ccccc3C

InChI

1S/C23H23N3/c1-16-10-5-7-12-20(16)24-18(3)22-14-9-15-23(26-22)19(4)25-21-13-8-6-11-17(21)2/h5-15H,1-4H3/b24-18+,25-19+

InChI key

FWPJMGBMVWKPTM-SIHVKLMXSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 4

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service