Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690589

Sigma-Aldrich

(R)-(+)-α-Methylvaline

99%, ≥98.5% (TLC)

Synonym(s):

α-Me-D-Val-OH, α-Methyl-D-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (TLC)
99%

form

powder

optical activity

[α]/D +3.5±0.5°, c = 1% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

impurities

≤0.2% water

application(s)

peptide synthesis

SMILES string

CC(C)[C@@](C)(N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)6(3,7)5(8)9/h4H,7H2,1-3H3,(H,8,9)/t6-/m1/s1

InChI key

GPYTYOMSQHBYTK-ZCFIWIBFSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service