Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690538

Sigma-Aldrich

3-Aminocrotononitrile

Arxada quality, ≥96.0% (based on DM, GC, KF)

Synonym(s):

3-Amino-2-butenenitrile, 3-Iminobutyronitrile, Diacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH3)=CHCN
CAS Number:
Molecular Weight:
82.10
Beilstein:
1719815
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (based on DM, GC, KF)

form

flakes

quality

Arxada quality

manufacturer/tradename

Arxada AG

storage temp.

2-8°C

SMILES string

C\C(N)=C/C#N

InChI

1S/C4H6N2/c1-4(6)2-3-5/h2H,6H2,1H3/b4-2+

InChI key

DELJOESCKJGFML-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service