Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690325

Sigma-Aldrich

D-Valinamide hydrochloride

97%

Synonym(s):

(R)-2-Amino-3-methylbutanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O · HCl
CAS Number:
Molecular Weight:
152.62
Beilstein:
5759926
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

96.5-103.5% (AT)
97%

form

powder

optical activity

[α]/D -26±1.5°, c = 1% in H2O

SMILES string

Cl.CC(C)[C@@H](N)C(N)=O

InChI

1S/C5H12N2O.ClH/c1-3(2)4(6)5(7)8;/h3-4H,6H2,1-2H3,(H2,7,8);1H/t4-;/m1./s1

InChI key

XFCNYSGKNAWXFL-PGMHMLKASA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Hasegawa et al.
ChemPlusChem, 85(2), 294-300 (2020-01-23)
Ligands based on 2,2'-bipyridine and valinamide moieties induce circularly polarized luminescence in their europium complexes. Both the R and S enantiomers of the complexes were successfully obtained. Single-crystal X-ray analysis of the racemic crystal confirmed that the ligand is coordinated
Takahiro Doi et al.
Forensic toxicology, 36(1), 51-60 (2018-01-26)
Recently, carboxamide-type synthetic cannabinoids have been distributed globally as new psychoactive substances (NPS). Some of these compounds possess asymmetric carbon, which is derived from an amide moiety composed of amino acid derivatives (i.e., amides or esters of amino acids). In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service