Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

689750

Sigma-Aldrich

3-Methyl-5-pyrazolone

99.0-100.3 % (w/w) (T)

Synonym(s):

2,4-Dihydro-5-methyl-3H-pyrazol-3-one, 3-Methyl-2-pyrazolin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

description

Lonza quality

form

powder

manufacturer/tradename

Lonza Ltd

concentration

99.0-100.3 % (w/w) (T)

mp

217.0-223.0 °C

SMILES string

O=C1CC(C)=NN1

InChI

1S/C4H6N2O/c1-3-2-4(7)6-5-3/h2H2,1H3,(H,6,7)

InChI key

NHLAPJMCARJFOG-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service