Skip to Content
MilliporeSigma
All Photos(1)

Documents

689521

Sigma-Aldrich

Dehydroacetic acid sodium salt

Arxada quality, 99.0-100.3 % (w/w) (T)

Synonym(s):

2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid δ-lactone sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NaO4
CAS Number:
Molecular Weight:
190.13
Beilstein:
4893653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

quality

Arxada quality

manufacturer/tradename

Arxada AG

concentration

99.0-100.3 % (w/w) (T)

impurities

≤0.5 % (w/w) water (Karl Fischer)

mp

~295 °C (dec.)

SMILES string

[Na+].CC(=O)C1=C([O-])C=C(C)OC1=O

InChI

1S/C8H8O4.Na/c1-4-3-6(10)7(5(2)9)8(11)12-4;/h3,10H,1-2H3;/q;+1/p-1

InChI key

ZPNRBQVNNIDJHX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Hata et al.
British journal of obstetrics and gynaecology, 105(3), 360-361 (1998-04-09)
We studied the effect of dehydroepiandrosterone sulphate (DHAS) on placental blood flow in 11 women with an uncomplicated pregnancy between 37 and 39 weeks of gestation. Power Doppler enhancement of the placenta was performed before and 60 minutes after the
Allergic contact dermatitis caused by sodium dehydroacetate, not hyaluronic acid, in Ialuset® cream.
Brigitte Milpied et al.
Contact dermatitis, 65(6), 359-361 (2011-11-15)
Huizhen Zhang et al.
Wei sheng yan jiu = Journal of hygiene research, 36(6), 748-750 (2008-02-29)
To estabish a high performance liquid chromatographic (HPLC) method for the determination of dehydroaetic acid in food determinated. Separation was carried out on a column of C18 (51 microm, 250 x 4.60mm) at the temperature of 25 degrees C with
Sodium dehydroacetate: an emerging allergen.
Caterina Foti et al.
Dermatitis : contact, atopic, occupational, drug, 23(5), 243-243 (2012-09-27)
Yansheng Wang et al.
Journal of food science, 75(6), M383-M388 (2010-08-21)
The peptide mixture from housefly pupae has broad spectrum antimicrobial activity but has not previously been reported as a food preservative. In this study, the preservation effects of a housefly pupae peptide mixture, nisin, and sodium dehydroacetate (DHA-S) on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service