Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688894

Sigma-Aldrich

Methyl 3-aminocrotonate

Arxada quality, 98.0-100.3 % (w/w) (GC)

Synonym(s):

Methyl 3-amino-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(NH2)=CHCOOCH3
CAS Number:
Molecular Weight:
115.13
Beilstein:
956592
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

crystals

quality

Arxada quality

manufacturer/tradename

Arxada AG

concentration

98.0-100.3 % (w/w) (GC)

impurities

≤1.0 % (w/w) water (Karl Fischer)

mp

81-83 °C (lit.)
81.0-84.0 °C

SMILES string

COC(=O)\C=C(\C)N

InChI

1S/C5H9NO2/c1-4(6)3-5(7)8-2/h3H,6H2,1-2H3/b4-3+

InChI key

XKORCTIIRYKLLG-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service