Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688800

Sigma-Aldrich

Dicyclohexyl-{2-sulfo-9-[3-(4-sulfo-phenyl)propyl]-9-fluorenyl}phosphonium-hydrogensulfate

95%

Synonym(s):

cataCXium® FSulf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H43O10PS3
CAS Number:
Molecular Weight:
738.87
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Asymmetric synthesis

functional group

phosphine

SMILES string

OS([O-])(=O)=O.OS(=O)(=O)c1ccc(CCCC4([PH+](C2CCCCC2)C3CCCCC3)c5ccccc5-c6ccc(cc46)S(O)(=O)=O)cc1

InChI

1S/C34H41O6PS2.H2O4S/c35-42(36,37)28-19-17-25(18-20-28)10-9-23-34(41(26-11-3-1-4-12-26)27-13-5-2-6-14-27)32-16-8-7-15-30(32)31-22-21-29(24-33(31)34)43(38,39)40;1-5(2,3)4/h7-8,15-22,24,26-27H,1-6,9-14,23H2,(H,35,36,37)(H,38,39,40);(H2,1,2,3,4)

InChI key

HNJNDGZFLWQSEG-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Legal Information

cataCXium is a registered trademark of Umicore AG & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous cross-coupling: highly efficient Suzuki-Miyaura coupling of N-heteroaryl halides and N-heteroarylboronic acids.
Fleckenstein, C. A.; Plenio, H.
Green Chemistry, 9, 1287-1287 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service