Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688517

Sigma-Aldrich

2-Chloroquinoline-3-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO2
CAS Number:
Molecular Weight:
207.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

199-240 °C

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1cc2ccccc2nc1Cl

InChI

1S/C10H6ClNO2/c11-9-7(10(13)14)5-6-3-1-2-4-8(6)12-9/h1-5H,(H,13,14)

InChI key

XFSORZYTTCOBFN-UHFFFAOYSA-N

Application

Useful quinolinecarboxylic acid for quinoline chemistry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Heterocyclic Chemistry, 26, 1589-1589 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service