Skip to Content
MilliporeSigma
All Photos(1)

Documents

688312

Sigma-Aldrich

2-Bromo-3-dodecylthiophene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27BrS
CAS Number:
Molecular Weight:
331.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

refractive index

n20/D 1.509

density

1.105 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCc1ccsc1Br

InChI

1S/C16H27BrS/c1-2-3-4-5-6-7-8-9-10-11-12-15-13-14-18-16(15)17/h13-14H,2-12H2,1H3

InChI key

IMILVTHOOISGRW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service