Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687944

Sigma-Aldrich

Ruthenium Metathesis Catalysts Kit I

Synonym(s):

Ruthenium Metathesis Catalysts

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.22

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

storage temp.

2-8°C

Application

Metathesis: Ruthenium-Based Metathesis Catalysts
Ruthenium metathesis catalysts kit I consists of 9 samples of Grubbs 1st and 2nd generation catalysts. These catalysts have applications in ring-closing and ring-opening metathesis, cross-metathesis, ring-opening metathesis polymerization (ROMP) and enyne metathesis.

Legal Information

Each individual catalyst contained in this product is subject to one or more US patents and associated foreign equivalents thereof. This product includes a limited one-time, non-exclusive, non-transferable license to use the Product for research purposes only. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the rights of others. For questions, please contact us at [email protected]
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Kit Components Also Available Separately

Product No.
Description
SDS

  • 579726Grubbs Catalyst® M102, Umicore, 97% 1 gSDS

  • 569747Grubbs Catalyst® M204, Umicore 100 mgSDS

  • 569755Hoveyda-Grubbs Catalyst® M720, Umicore, 97% 100 mgSDS

  • 577944Hoveyda-Grubbs Catalyst® M700, Umicore 100 mgSDS

  • 682284Grubbs Catalyst® M205, Umicore 100 mgSDS

  • 682330Grubbs Catalyst® M300 100 mgSDS

  • 682365Grubbs Catalyst® M207, Umicore 100 mgSDS

  • 682373Hoveyda-Grubbs Catalyst® M721, Umicore 100 mgSDS

  • 682381Grubbs Catalyst® M360 100 mgSDS

See All (9)

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general model for selectivity in olefin cross metathesis
Chatterjee, AK et al.
Journal of the American Chemical Society, 125(37), 11360-11370 (2003)
Enyne metathesis (enyne bond reorganization)
Diver, ST and Giessert, A
Chemical Reviews, 104(3), 1317-1382 (2004)
Controlled Living Ring-Opening-Metathesis Polymerization by a Fast-Initiating Ruthenium Catalyst
Choi, T and Grubbs, RH
Angewandte Chemie (International ed. in English), 42(15), 1743-1746 (2003)
The total synthesis of (+)-migrastatin.
Gaul C, et al.
Journal of the American Chemical Society, 125(20), 6042-6043 (2003)
Schrodi, Y.; Pederson, R.L.
Aldrichimica Acta, 40, 45-45 (2007)

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service