Skip to Content
MilliporeSigma
All Photos(1)

Documents

687928

Sigma-Aldrich

2-Ethylbenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4COCl
CAS Number:
Molecular Weight:
168.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.545

density

1.127 g/mL at 25 °C

SMILES string

CCc1ccccc1C(Cl)=O

InChI

1S/C9H9ClO/c1-2-7-5-3-4-6-8(7)9(10)11/h3-6H,2H2,1H3

InChI key

PJGFNNXYKMSCCU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service