Skip to Content
MilliporeSigma
All Photos(4)

Documents

687812

Sigma-Aldrich

Carbon nanotube array, multi-walled

vertically aligned on copper wafer substrate

Synonym(s):

CNT array, MWCNT array

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

material

copper (wafer)

Assay

>99.9% carbon basis

form

(1cm X 1cm X 0.05cm high conductivity low-oxygen copper substrate)

composition

Carbon, >99.9% (structured (sp2) carbon. <0.1% amorphous (sp3) carbon.)

diam. × L

100 nm × 30 μm , ±10%

surface coverage

surface coverage ~2 × 109 CNT/cm2 ~20/μm2

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Multi walled carbon nanotubes (MWNTs, CNTs) was prepared by plasma enhanced chemical vapor deposition (PECVD). The CNTs formed can be formed in bundles over a catalyst support or in patterns over substrates.1 Each MWCNT is tipped with nickel-catalyst particles with diameters equal to the diameter of the nanotube.

Application

High quality CNT arrays suitable for nanoelectronics

Features and Benefits

Each MWCNT is tipped with nickel-carbide particle with diameter equal to diameter of the nanotube.

Packaging

Packaged in Vacuum Release (VR) Trays. See Technical bulletin AL-237.

Preparation Note

Plasma-Enhanced Chemical Vapor Deposition (PECVD) Method

Legal Information

Vacuum Release is a trademark of Gel-Pak

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service